Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCAGGAATGACAAGATGCAGGAA[G/T]GGCCTCCTCTTCTCACACCTGTCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611580 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EDDM3A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
EDDM3A - epididymal protein 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006683.4 | Intron | NP_006674.2 | ||||
XM_017020934.1 | Intron | XP_016876423.1 |
Set Membership: |
HapMap Validated |