Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610858 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf166 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C14orf166 - chromosome 14 open reading frame 166 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016039.2 | Intron | NP_057123.1 |