Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACCGGGCAAATAGATTCGAGTA[C/T]TTATTAAAGCAGACAGAACTTTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603375 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMARCA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SMARCA5 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003601.3 | 744 | Silent Mutation | TAC,TAT | Y,Y 94 | NP_003592.3 |
SMARCA5-AS1 - SMARCA5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |