Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATTTACCTCTTGGCTGTTTAGT[C/T]TCGGGCAAATTACCTGTCTGCTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606676 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC75A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LRRC75A - leucine rich repeat containing 75A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC75A-AS1 - LRRC75A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD49A - small nucleolar RNA, C/D box 49A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD49B - small nucleolar RNA, C/D box 49B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD65 - small nucleolar RNA, C/D box 65 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRPV2 - transient receptor potential cation channel subfamily V member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |