Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGACTTTCCATTTAGCGAAAAAG[A/G]CAGATGTTGCGCAAAAAACAAATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609482 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AGR3 - anterior gradient 3, protein disulphide isomerase family member | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176813.4 | Intron | NP_789783.1 | ||||
XM_005249627.2 | Intron | XP_005249684.1 | ||||
XM_011515152.2 | Intron | XP_011513454.1 |
Set Membership: |
HapMap |