Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM178B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM178B - family with sequence similarity 178 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122646.2 | Intron | NP_001116118.2 | ||||
NM_001172667.1 | Intron | NP_001166138.1 | ||||
NM_016490.4 | Intron | NP_057574.2 | ||||
XM_011511294.2 | Intron | XP_011509596.1 | ||||
XM_011511297.2 | Intron | XP_011509599.1 | ||||
XM_017004263.1 | Intron | XP_016859752.1 | ||||
XM_017004264.1 | Intron | XP_016859753.1 | ||||
XM_017004265.1 | Intron | XP_016859754.1 | ||||
XM_017004266.1 | Intron | XP_016859755.1 | ||||
XM_017004267.1 | Intron | XP_016859756.1 | ||||
XM_017004268.1 | Intron | XP_016859757.1 |