Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTAGGTCACATTTCTGTGAAGAA[C/T]AGCATCATCGAAAACCCTTGCTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP10D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP10D - ATPase phospholipid transporting 10D (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020453.3 | Intron | NP_065186.3 | ||||
XM_005248119.4 | Intron | XP_005248176.1 | ||||
XM_005248120.4 | Intron | XP_005248177.1 | ||||
XM_011513722.2 | Intron | XP_011512024.1 | ||||
XM_017008472.1 | Intron | XP_016863961.1 |