Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601296 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MUSK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MUSK - muscle associated receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166280.1 | Intron | NP_001159752.1 | ||||
NM_001166281.1 | Intron | NP_001159753.1 | ||||
NM_005592.3 | Intron | NP_005583.1 | ||||
XM_005251994.3 | Intron | XP_005252051.1 | ||||
XM_005251995.3 | Intron | XP_005252052.1 | ||||
XM_005251996.3 | Intron | XP_005252053.1 | ||||
XM_011518708.2 | Intron | XP_011517010.1 | ||||
XM_017014734.1 | Intron | XP_016870223.1 |
Set Membership: |
HapMap Validated |