Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTAGATCAGCATTATTAAAGAAA[C/T]GGTTCAACTTTGTTTCTTCCCTTAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 112203 MIM: 615534 | |||||||||||||||||||||||
Literature Links: |
CD80 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CD80 - CD80 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005191.3 | 4811 | UTR 3 | NP_005182.1 | |||
XM_011513327.2 | 4811 | UTR 3 | XP_011511629.1 | |||
XM_017007520.1 | 4811 | Intron | XP_016863009.1 |
TIMMDC1 - translocase of inner mitochondrial membrane domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016589.3 | 4811 | Intron | NP_057673.2 | |||
XM_017006556.1 | 4811 | Intron | XP_016862045.1 |