Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602886 MIM: 610450 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR39 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GPR39 - G protein-coupled receptor 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001508.2 | 1786 | Intron | NP_001499.1 | |||
XM_011511021.2 | 1786 | Intron | XP_011509323.1 |
LYPD1 - LY6/PLAUR domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077427.3 | 1786 | UTR 3 | NP_001070895.1 | |||
NM_001321234.1 | 1786 | UTR 3 | NP_001308163.1 | |||
NM_001321235.1 | 1786 | UTR 3 | NP_001308164.1 | |||
NM_144586.6 | 1786 | UTR 3 | NP_653187.3 | |||
XM_011510546.2 | 1786 | UTR 3 | XP_011508848.1 |
Set Membership: |
HapMap Validated |