Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610668 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INSC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INSC - inscuteable homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031853.4 | Intron | NP_001027024.3 | ||||
NM_001042536.2 | Intron | NP_001036001.1 | ||||
NM_001278313.1 | Intron | NP_001265242.1 | ||||
NM_001278314.1 | Intron | NP_001265243.1 | ||||
NM_001278315.1 | Intron | NP_001265244.1 | ||||
NM_001278316.1 | Intron | NP_001265245.1 | ||||
XM_006718227.3 | Intron | XP_006718290.1 | ||||
XM_011520085.1 | Intron | XP_011518387.1 | ||||
XM_011520086.1 | Intron | XP_011518388.1 | ||||
XM_017017697.1 | Intron | XP_016873186.1 | ||||
XM_017017698.1 | Intron | XP_016873187.1 |