Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGBL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AGBL3 - ATP/GTP binding protein like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178563.3 | 540 | Intron | NP_848658.3 | |||
XM_011516122.2 | 540 | Intron | XP_011514424.1 | |||
XM_017012136.1 | 540 | UTR 5 | XP_016867625.1 | |||
XM_017012137.1 | 540 | UTR 5 | XP_016867626.1 | |||
XM_017012138.1 | 540 | Intron | XP_016867627.1 | |||
XM_017012139.1 | 540 | Intron | XP_016867628.1 | |||
XM_017012140.1 | 540 | Intron | XP_016867629.1 | |||
XM_017012141.1 | 540 | UTR 5 | XP_016867630.1 | |||
XM_017012142.1 | 540 | Intron | XP_016867631.1 | |||
XM_017012143.1 | 540 | Intron | XP_016867632.1 |
Set Membership: |
HapMap Validated |