Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTCCCCAGGCTGGGGCCTGGGC[C/G]AGTAAAGCATGCCCCTCCTACTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612545 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ABHD11 - abhydrolase domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145364.2 | 1204 | Intron | NP_001138836.1 | |||
NM_001301058.1 | 1204 | Intron | NP_001287987.1 | |||
NM_001321382.1 | 1204 | Intron | NP_001308311.1 | |||
NM_001321383.1 | 1204 | UTR 3 | NP_001308312.1 | |||
NM_148912.3 | 1204 | Intron | NP_683710.1 | |||
NM_148913.3 | 1204 | Intron | NP_683711.1 | |||
XM_006716144.2 | 1204 | Intron | XP_006716207.1 | |||
XM_011516590.2 | 1204 | Intron | XP_011514892.1 | |||
XM_011516591.1 | 1204 | Intron | XP_011514893.1 |
ABHD11-AS1 - ABHD11 antisense RNA 1 (tail to tail) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |