Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGATATGAGATAGCAAAGTATA[C/T]GTACAGTACCTGGAACCATAATCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608555 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MTX2 - metaxin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006635.2 | Intron | NP_001006636.1 | ||||
NM_001319097.1 | Intron | NP_001306026.1 | ||||
NM_001319098.1 | Intron | NP_001306027.1 | ||||
NM_006554.4 | Intron | NP_006545.1 | ||||
XM_011510497.2 | Intron | XP_011508799.1 |