Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTGAGGAATCACTGTCATCACT[A/G]TAAGGTAAGTCTTTTGAGCTAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607710 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM189A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM189A2 - family with sequence similarity 189 member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127608.1 | Intron | NP_001121080.1 | ||||
NM_004816.3 | Intron | NP_004807.3 | ||||
XM_005252307.4 | Intron | XP_005252364.1 | ||||
XM_017015324.1 | Intron | XP_016870813.1 | ||||
XM_017015325.1 | Intron | XP_016870814.1 |