Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603722 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IKBKAP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IKBKAP - inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318360.1 | Intron | NP_001305289.1 | ||||
NM_003640.4 | Intron | NP_003631.2 | ||||
XM_011519136.1 | Intron | XP_011517438.1 | ||||
XM_011519137.1 | Intron | XP_011517439.1 | ||||
XM_017015238.1 | Intron | XP_016870727.1 |
Set Membership: |
HapMap |