Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAAAACTGTTATCATAAGTATAC[A/C]GTTGGATGACTTTTCATAAAGTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604593 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF5C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF5C - kinesin family member 5C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004522.2 | Intron | NP_004513.1 | ||||
XM_011511157.2 | Intron | XP_011509459.1 | ||||
XM_017004062.1 | Intron | XP_016859551.1 |
LOC101928553 - uncharacterized LOC101928553 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |