Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603337 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAH8 - dynein axonemal heavy chain 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206927.1 | Intron | NP_001193856.1 | ||||
XM_011514318.2 | Intron | XP_011512620.1 | ||||
XM_011514319.2 | Intron | XP_011512621.1 | ||||
XM_011514320.2 | Intron | XP_011512622.1 | ||||
XM_011514321.1 | Intron | XP_011512623.1 | ||||
XM_017010325.1 | Intron | XP_016865814.1 | ||||
XM_017010326.1 | Intron | XP_016865815.1 | ||||
XM_017010327.1 | Intron | XP_016865816.1 |