Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALPK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALPK2 - alpha kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052947.3 | 5852 | UTR 3 | NP_443179.3 | |||
XM_011525801.2 | 5852 | Intron | XP_011524103.1 | |||
XM_011525802.2 | 5852 | UTR 3 | XP_011524104.1 | |||
XM_017025535.1 | 5852 | Intron | XP_016881024.1 | |||
XM_017025536.1 | 5852 | Intron | XP_016881025.1 | |||
XM_017025537.1 | 5852 | Intron | XP_016881026.1 | |||
XM_017025538.1 | 5852 | UTR 3 | XP_016881027.1 | |||
XM_017025539.1 | 5852 | UTR 3 | XP_016881028.1 |
Set Membership: |
HapMap |