Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATCAGGCCTCCCAGAGCCTGGG[C/T]GTCAGCTGAGGACTCCCATGATCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615563 MIM: 604819 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6845 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR6845 - microRNA 6845 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRBP2 - nuclear receptor binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178564.3 | Intron | NP_848659.2 | ||||
XM_017013378.1 | Intron | XP_016868867.1 | ||||
XM_017013379.1 | Intron | XP_016868868.1 | ||||
XM_017013380.1 | Intron | XP_016868869.1 | ||||
XM_017013381.1 | Intron | XP_016868870.1 | ||||
XM_017013382.1 | Intron | XP_016868871.1 | ||||
XM_017013383.1 | Intron | XP_016868872.1 |
PUF60 - poly(U) binding splicing factor 60 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |