Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAGAAAGCAGGCTTAAAATTTTA[A/G]TAGGTAGGATTTAGATGAGACAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
57 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611347 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
INTS3 - integrator complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324475.1 | Intron | NP_001311404.1 | ||||
NM_023015.4 | Intron | NP_075391.3 | ||||
XM_005245459.2 | Intron | XP_005245516.1 | ||||
XM_005245461.2 | Intron | XP_005245518.1 | ||||
XM_006711490.3 | Intron | XP_006711553.1 | ||||
XM_011509906.2 | Intron | XP_011508208.1 | ||||
XM_017002132.1 | Intron | XP_016857621.1 |