Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTGTAAGGAAGGTTTGCTCGGC[A/T]GTTCCCCTGATAGGGCCTAAGGGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604374 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TESMIN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TESMIN - testis expressed metallothionein like protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039656.1 | Intron | NP_001034745.1 | ||||
NM_004923.3 | Intron | NP_004914.2 | ||||
XM_011545402.1 | Intron | XP_011543704.1 | ||||
XM_011545403.2 | Intron | XP_011543705.1 | ||||
XM_017018588.1 | Intron | XP_016874077.1 |