Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 115435 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAPLN1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HAPLN1 - hyaluronan and proteoglycan link protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001884.3 | 2363 | UTR 3 | NP_001875.1 | |||
XM_011543168.2 | 2363 | UTR 3 | XP_011541470.1 | |||
XM_017009051.1 | 2363 | UTR 3 | XP_016864540.1 | |||
XM_017009052.1 | 2363 | UTR 3 | XP_016864541.1 | |||
XM_017009053.1 | 2363 | UTR 3 | XP_016864542.1 | |||
XM_017009054.1 | 2363 | UTR 3 | XP_016864543.1 |
Set Membership: |
HapMap |