Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
55 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601836 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIFAP3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIFAP3 - kinesin associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204514.1 | Intron | NP_001191443.1 | ||||
NM_001204516.1 | Intron | NP_001191445.1 | ||||
NM_001204517.1 | Intron | NP_001191446.1 | ||||
NM_014970.3 | Intron | NP_055785.2 | ||||
XM_005244970.1 | Intron | XP_005245027.1 | ||||
XM_011509307.2 | Intron | XP_011507609.2 |
Set Membership: |
HapMap JSNP |