Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGAGTTCATTTCTCAAGATAATG[T/C]CCAATTTGTCTGCATTTTCAGCAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613349 | ||||||||||||||||||||
Literature Links: |
OAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OAT - ornithine aminotransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000274.3 | Intron | NP_000265.1 | ||||
NM_001171814.1 | Intron | NP_001165285.1 | ||||
NM_001322965.1 | Intron | NP_001309894.1 | ||||
NM_001322966.1 | Intron | NP_001309895.1 | ||||
NM_001322967.1 | Intron | NP_001309896.1 | ||||
NM_001322968.1 | Intron | NP_001309897.1 | ||||
NM_001322969.1 | Intron | NP_001309898.1 | ||||
NM_001322970.1 | Intron | NP_001309899.1 | ||||
NM_001322971.1 | Intron | NP_001309900.1 | ||||
NM_001322974.1 | Intron | NP_001309903.1 | ||||
XM_017016279.1 | Intron | XP_016871768.1 |