Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601698 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRN2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PTPRN2 - protein tyrosine phosphatase, receptor type N2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308267.1 | 4417 | UTR 3 | NP_001295196.1 | |||
NM_001308268.1 | 4417 | UTR 3 | NP_001295197.1 | |||
NM_002847.4 | 4417 | UTR 3 | NP_002838.2 | |||
NM_130842.3 | 4417 | UTR 3 | NP_570857.2 | |||
NM_130843.3 | 4417 | UTR 3 | NP_570858.2 | |||
XM_011516446.1 | 4417 | Intron | XP_011514748.1 | |||
XM_011516447.2 | 4417 | Intron | XP_011514749.1 | |||
XM_011516448.2 | 4417 | Intron | XP_011514750.1 | |||
XM_011516449.2 | 4417 | Intron | XP_011514751.1 | |||
XM_017012475.1 | 4417 | UTR 3 | XP_016867964.1 | |||
XM_017012476.1 | 4417 | Intron | XP_016867965.1 |
Set Membership: |
HapMap Validated |