Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATTAGGCTTCCTGAGCTCGGTC[A/G]GTTTTTCCCTGTGATCCAGTCATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612676 | ||||||||||||||||||||
Literature Links: |
QDPR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
QDPR - quinoid dihydropteridine reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000320.2 | 748 | Nonsense Mutation | CGA,TGA | R,* 221 | NP_000311.2 | |
NM_001306140.1 | 748 | Nonsense Mutation | CGA,TGA | R,* 190 | NP_001293069.1 |