Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602465 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPRY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPRY1 - sprouty RTK signaling antagonist 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258038.1 | 568 | Intron | NP_001244967.1 | |||
NM_001258039.1 | 568 | Intron | NP_001244968.1 | |||
NM_005841.2 | 568 | Intron | NP_005832.1 | |||
NM_199327.2 | 568 | Intron | NP_955359.1 | |||
XM_005262686.1 | 568 | UTR 5 | XP_005262743.1 | |||
XM_017007635.1 | 568 | Intron | XP_016863124.1 |
Set Membership: |
HapMap |