Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615370 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKS6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKS6 - ankyrin repeat and sterile alpha motif domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173551.4 | 5122 | UTR 3 | NP_775822.3 | |||
XM_005251793.4 | 5122 | Intron | XP_005251850.1 | |||
XM_005251794.4 | 5122 | Intron | XP_005251851.1 | |||
XM_006716998.3 | 5122 | Intron | XP_006717061.1 | |||
XM_006716999.2 | 5122 | Intron | XP_006717062.1 | |||
XM_011518357.2 | 5122 | Intron | XP_011516659.1 | |||
XM_017014445.1 | 5122 | Intron | XP_016869934.1 | |||
XM_017014446.1 | 5122 | Intron | XP_016869935.1 | |||
XM_017014447.1 | 5122 | Intron | XP_016869936.1 |
Set Membership: |
HapMap |