Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
31 submissions
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 142702 | |||||||||||||||||||||||||||||
Literature Links: |
HTN3 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EUR
|
African American - Not Available | YRI (Yoruba)
|
||||||
EAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
SAS - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
AFR - Not Available | ||||||||
HTN3 - histatin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000200.2 | 239 | Missense Mutation | CAA,CGA | Q,R 41 | NP_000191.1 |