Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611018 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAIP2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PAIP2B - poly(A) binding protein interacting protein 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020459.1 | 2367 | UTR 3 | NP_065192.1 | |||
XM_005264310.3 | 2367 | UTR 3 | XP_005264367.1 | |||
XM_005264311.4 | 2367 | UTR 3 | XP_005264368.1 | |||
XM_011532842.2 | 2367 | UTR 3 | XP_011531144.1 | |||
XM_011532843.2 | 2367 | UTR 3 | XP_011531145.1 |