Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613713 MIM: 176895 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCID2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PCID2 - PCI domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127202.3 | 2342 | UTR 3 | NP_001120674.1 | |||
NM_001127203.3 | 2342 | UTR 3 | NP_001120675.1 | |||
NM_001258213.2 | 2342 | UTR 3 | NP_001245142.1 | |||
NM_001320655.1 | 2342 | UTR 3 | NP_001307584.1 | |||
NM_001320656.1 | 2342 | UTR 3 | NP_001307585.1 | |||
NM_001320657.1 | 2342 | UTR 3 | NP_001307586.1 | |||
NM_001320659.1 | 2342 | UTR 3 | NP_001307588.1 | |||
NM_001320660.1 | 2342 | UTR 3 | NP_001307589.1 | |||
NM_018386.4 | 2342 | UTR 3 | NP_060856.2 | |||
XM_005268325.3 | 2342 | Intron | XP_005268382.1 | |||
XM_017020664.1 | 2342 | UTR 3 | XP_016876153.1 | |||
XM_017020665.1 | 2342 | UTR 3 | XP_016876154.1 | |||
XM_017020666.1 | 2342 | UTR 3 | XP_016876155.1 | |||
XM_017020667.1 | 2342 | UTR 3 | XP_016876156.1 | |||
XM_017020668.1 | 2342 | UTR 3 | XP_016876157.1 | |||
XM_017020669.1 | 2342 | UTR 3 | XP_016876158.1 | |||
XM_017020670.1 | 2342 | UTR 3 | XP_016876159.1 | |||
XM_017020671.1 | 2342 | UTR 3 | XP_016876160.1 | |||
XM_017020672.1 | 2342 | UTR 3 | XP_016876161.1 |
PROZ - protein Z, vitamin K dependent plasma glycoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |