Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTGTAAGCATACCAAAATAATA[A/C]AGTATAGCCCACGTATGAGCCAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609216 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371998 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105371998 - uncharacterized LOC105371998 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPIRE1 - spire type actin nucleation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128626.1 | 3888 | UTR 3 | NP_001122098.1 | |||
NM_001128627.1 | 3888 | UTR 3 | NP_001122099.1 | |||
NM_020148.2 | 3888 | UTR 3 | NP_064533.3 | |||
XM_005258122.4 | 3888 | UTR 3 | XP_005258179.1 | |||
XM_011525701.1 | 3888 | UTR 3 | XP_011524003.1 | |||
XM_011525702.1 | 3888 | UTR 3 | XP_011524004.1 | |||
XM_011525703.2 | 3888 | Intron | XP_011524005.1 |
Set Membership: |
HapMap |