Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||
Literature Links: |
JMJD4 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
JMJD4 - jumonji domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161465.1 | 367 | Missense Mutation | AAG,GAG | K,E 407 | NP_001154937.1 | |
NM_023007.2 | 367 | Missense Mutation | AAG,GAG | K,E 423 | NP_075383.2 | |
XM_011544262.2 | 367 | Missense Mutation | AAG,GAG | K,E 374 | XP_011542564.1 |
SNAP47 - synaptosome associated protein 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323930.1 | 367 | Intron | NP_001310859.1 | |||
NM_001323931.1 | 367 | Intron | NP_001310860.1 | |||
NM_001323932.1 | 367 | Intron | NP_001310861.1 | |||
NM_001323933.1 | 367 | Intron | NP_001310862.1 | |||
NM_001323934.1 | 367 | Intron | NP_001310863.1 | |||
NM_001323935.1 | 367 | Intron | NP_001310864.1 | |||
NM_053052.3 | 367 | Intron | NP_444280.2 | |||
XM_006711734.3 | 367 | Intron | XP_006711797.1 | |||
XM_017000231.1 | 367 | UTR 5 | XP_016855720.1 | |||
XM_017000232.1 | 367 | UTR 5 | XP_016855721.1 |
Set Membership: |
HapMap |