Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603743 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
APOL1 - apolipoprotein L1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136540.1 | Intron | NP_001130012.1 | ||||
NM_001136541.1 | Intron | NP_001130013.1 | ||||
NM_003661.3 | Intron | NP_003652.2 | ||||
NM_145343.2 | Intron | NP_663318.1 | ||||
XM_005261796.3 | Intron | XP_005261853.1 | ||||
XM_011530478.2 | Intron | XP_011528780.1 |
Set Membership: |
HapMap |