Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf70 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C2orf70 - chromosome 2 open reading frame 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105519.2 | Intron | NP_001098989.1 | ||||
NM_001322426.1 | Intron | NP_001309355.1 | ||||
XM_011532810.2 | Intron | XP_011531112.1 | ||||
XM_011532811.2 | Intron | XP_011531113.1 | ||||
XM_017003962.1 | Intron | XP_016859451.1 | ||||
XM_017003963.1 | Intron | XP_016859452.1 | ||||
XM_017003964.1 | Intron | XP_016859453.1 | ||||
XM_017003965.1 | Intron | XP_016859454.1 | ||||
XM_017003966.1 | Intron | XP_016859455.1 |