Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607162 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST8SIA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ST8SIA5 - ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307986.1 | Intron | NP_001294915.1 | ||||
NM_001307987.1 | Intron | NP_001294916.1 | ||||
NM_013305.5 | Intron | NP_037437.2 | ||||
XM_017025715.1 | Intron | XP_016881204.1 | ||||
XM_017025716.1 | Intron | XP_016881205.1 |
Set Membership: |
HapMap |