Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 168470 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTHLH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTHLH - parathyroid hormone like hormone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002820.2 | 1167 | UTR 3 | NP_002811.1 | |||
NM_198964.1 | 1167 | UTR 3 | NP_945315.1 | |||
NM_198965.1 | 1167 | Intron | NP_945316.1 | |||
NM_198966.1 | 1167 | Intron | NP_945317.1 | |||
XM_011520774.2 | 1167 | Intron | XP_011519076.1 | |||
XM_011520775.2 | 1167 | Intron | XP_011519077.1 | |||
XM_017019674.1 | 1167 | Intron | XP_016875163.1 | |||
XM_017019675.1 | 1167 | Intron | XP_016875164.1 |
Set Membership: |
Validated |