Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXD1 - exonuclease 3'-5' domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286441.1 | 1566 | UTR 3 | NP_001273370.1 | |||
NM_152596.3 | 1566 | UTR 3 | NP_689809.2 | |||
XM_006720415.3 | 1566 | UTR 3 | XP_006720478.1 | |||
XM_011521298.2 | 1566 | UTR 3 | XP_011519600.1 | |||
XM_011521299.2 | 1566 | UTR 3 | XP_011519601.1 | |||
XM_011521300.2 | 1566 | UTR 3 | XP_011519602.1 | |||
XM_011521301.2 | 1566 | UTR 3 | XP_011519603.1 | |||
XM_011521302.2 | 1566 | UTR 3 | XP_011519604.1 | |||
XM_017021970.1 | 1566 | UTR 3 | XP_016877459.1 | |||
XM_017021971.1 | 1566 | UTR 3 | XP_016877460.1 |