Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609977 MIM: 611123 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDCA8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDCA8 - cell division cycle associated 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EPHA10 - EPH receptor A10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099439.1 | 4824 | UTR 3 | NP_001092909.1 | |||
NM_173641.2 | 4824 | Intron | NP_775912.2 | |||
XM_017001080.1 | 4824 | Intron | XP_016856569.1 | |||
XM_017001081.1 | 4824 | Intron | XP_016856570.1 |
Set Membership: |
HapMap |