Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604599 MIM: 606107 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EHMT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EHMT2 - euchromatic histone lysine methyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289413.1 | Intron | NP_001276342.1 | ||||
NM_001318833.1 | Intron | NP_001305762.1 | ||||
NM_006709.4 | Intron | NP_006700.3 | ||||
NM_025256.6 | Intron | NP_079532.5 | ||||
XM_005248824.3 | Intron | XP_005248881.1 | ||||
XM_006714974.1 | Intron | XP_006715037.1 | ||||
XM_006714975.1 | Intron | XP_006715038.1 | ||||
XM_006714976.1 | Intron | XP_006715039.1 | ||||
XM_017010202.1 | Intron | XP_016865691.1 |
SLC44A4 - solute carrier family 44 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |