Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615680 MIM: 147678 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARD16 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CARD16 - caspase recruitment domain family member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017534.1 | 590 | Missense Mutation | AAC,ATC | N,I 167 | NP_001017534.1 | |
NM_052889.2 | 590 | UTR 3 | NP_443121.1 | |||
XM_011542583.2 | 590 | Missense Mutation | AAC,ATC | N,I 235 | XP_011540885.1 |
CASP1 - caspase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |