Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615256 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AMN1 - antagonist of mitotic exit network 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113402.1 | 4503 | UTR 3 | NP_001106873.1 | |||
NM_001278411.1 | 4503 | UTR 3 | NP_001265340.1 | |||
NM_001278412.1 | 4503 | UTR 3 | NP_001265341.1 | |||
XM_006719054.3 | 4503 | UTR 3 | XP_006719117.1 | |||
XM_011520604.2 | 4503 | UTR 3 | XP_011518906.1 | |||
XM_017018963.1 | 4503 | UTR 3 | XP_016874452.1 | |||
XM_017018964.1 | 4503 | UTR 3 | XP_016874453.1 | |||
XM_017018965.1 | 4503 | UTR 3 | XP_016874454.1 |
ETFBKMT - electron transfer flavoprotein beta subunit lysine methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135863.1 | 4503 | Intron | NP_001129335.1 | |||
NM_001135864.1 | 4503 | Intron | NP_001129336.1 | |||
NM_173802.3 | 4503 | Intron | NP_776163.1 |
Set Membership: |
HapMap Validated |