Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602647 MIM: 602946 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6514 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR6514 - microRNA 6514 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6748 - microRNA 6748 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NXF1 - nuclear RNA export factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081491.1 | Intron | NP_001074960.1 | ||||
NM_006362.4 | Intron | NP_006353.2 |
TAF6L - TATA-box binding protein associated factor 6 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM179B - transmembrane protein 179B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199337.2 | Intron | NP_955369.1 | ||||
XM_005273982.3 | Intron | XP_005274039.3 |
TMEM223 - transmembrane protein 223 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080501.2 | Intron | NP_001073970.1 |
Set Membership: |
HapMap |