Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606879 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHGDH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PHGDH - phosphoglycerate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006623.3 | 147 | UTR 5 | NP_006614.2 | |||
XM_011541226.1 | 147 | UTR 5 | XP_011539528.1 | |||
XM_011541227.2 | 147 | Intron | XP_011539529.1 | |||
XM_011541228.1 | 147 | Intron | XP_011539530.1 | |||
XM_011541231.2 | 147 | Intron | XP_011539533.1 |
Set Membership: |
HapMap |