Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608167 MIM: 610224 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNT1 - potassium sodium-activated channel subfamily T member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272003.1 | 2296 | Intron | NP_001258932.1 | |||
NM_020822.2 | 2296 | Intron | NP_065873.2 | |||
XM_011518877.2 | 2296 | UTR 5 | XP_011517179.1 | |||
XM_011518878.2 | 2296 | UTR 5 | XP_011517180.1 | |||
XM_011518879.2 | 2296 | UTR 5 | XP_011517181.1 | |||
XM_011518880.1 | 2296 | Intron | XP_011517182.1 | |||
XM_011518881.2 | 2296 | Intron | XP_011517183.1 | |||
XM_017014931.1 | 2296 | Intron | XP_016870420.1 | |||
XM_017014932.1 | 2296 | Intron | XP_016870421.1 | |||
XM_017014933.1 | 2296 | Intron | XP_016870422.1 |
SOHLH1 - spermatogenesis and oogenesis specific basic helix-loop-helix 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012415.2 | 2296 | Intron | NP_001012415.2 | |||
NM_001101677.1 | 2296 | Intron | NP_001095147.1 |
Set Membership: |
HapMap |