Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608167 MIM: 610224 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNT1 - potassium sodium-activated channel subfamily T member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOHLH1 - spermatogenesis and oogenesis specific basic helix-loop-helix 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012415.2 | 3024 | Intron | NP_001012415.2 | |||
NM_001101677.1 | 3024 | Intron | NP_001095147.1 | |||
XM_005263403.3 | 3024 | UTR 3 | XP_005263460.1 | |||
XM_006717109.3 | 3024 | UTR 3 | XP_006717172.1 | |||
XM_011518698.2 | 3024 | UTR 3 | XP_011517000.1 |