Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609312 MIM: 604455 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBH PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
DBH - dopamine beta-hydroxylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DBH-AS1 - DBH antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SARDH - sarcosine dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134707.1 | Intron | NP_001128179.1 | ||||
NM_007101.3 | Intron | NP_009032.2 | ||||
XM_017014367.1 | Intron | XP_016869856.1 | ||||
XM_017014368.1 | Intron | XP_016869857.1 |
Set Membership: |
HapMap |