Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGAAGGCTATACTGGGGCATGA[A/C]CACCAGGAGGCCAGGGATCACTGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616210 MIM: 231675 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf46 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
C4orf46 - chromosome 4 open reading frame 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008393.3 | 2380 | UTR 3 | NP_001008394.1 |
ETFDH - electron transfer flavoprotein dehydrogenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |