Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCCCTACCCTTGAAAATGGATG[C/G]TCTAATTTCTTCCTGTCCCCAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609540 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAJA4 - DnaJ heat shock protein family (Hsp40) member A4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR61 - WD repeat domain 61 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303247.1 | Intron | NP_001290176.1 | ||||
NM_001303248.1 | Intron | NP_001290177.1 | ||||
NM_025234.2 | Intron | NP_079510.1 | ||||
XM_011522094.1 | Intron | XP_011520396.1 | ||||
XM_017022637.1 | Intron | XP_016878126.1 |